Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents

Catalog No. p-29016 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP153-500 500 g Poly Bottle
BP153-1 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP153-500
Only null left
Add to Cart

For electrophoresis, molecular biology, and cell culture applications | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

For electrophoresis, molecular biology, and cell culture applications

Tris Hydrochloride is a buffering medium for electrophoresis, molecular biology, and cell culture applications. It may be combined with Tris Base to simplify Tris buffer preparation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane Hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Absorbance 0.05 max. (1M solution) at 280nm
Melting Point 150°C
Color White
pH 3.5 to 5.0
Physical Form Solid
Quantity 500 g
DNase DNase free
Identification Pass Test
Assay Percent Range ≥99 %
Linear Formula H2NC(CH2OH)3·HCl
Packaging Poly Bottle
Protease Protease free
Formula Weight 157.6
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Hydrochloride
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage Keep in a dry, cool and well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.