Learn More
Alkali Scientific TRIS Hydrochloride [Tris(hydroxymethyl) aminomethane HCl], 100 G
SDP
Catalog No. 5000659750
Encompass_Preferred
TRIS Hydrochloride [Tris (Hydroxymethyl) Aminomethane HCl], 100g, is a biochemical reagent used for buffer preparation and pH control in a variety of laboratory applications. This hydrochloride salt of Tris is often used in protein purification, nucleic acid analysis, and molecular biology experiments that require a stable acidic environment. The 100g size is suitable for laboratories requiring smaller quantities of Tris Hydrochloride for specific experiments. This reagent is commonly used in conjunction with other buffer systems to maintain the pH of reaction mixtures in biochemical assays, electrophoresis, and enzyme catalysis studies. The high-purity formulation ensures minimal impurities and excellent performance in sensitive experiments.
Catalog No. 50-006-59750 Supplier Alkali Scientific Supplier No. ISH001CP
TRIS Hydrochloride [Tris (Hydroxymethyl) Aminomethane HCl], 100g, is a biochemical reagent used for buffer preparation and pH control in a variety of laboratory applications. This hydrochloride salt of Tris is often used in protein purification, nucleic acid analysis, and molecular biology experiments that require a stable acidic environment. The 100g size is suitable for laboratories requiring smaller quantities of Tris Hydrochloride for specific experiments. This reagent is commonly used in conjunction with other buffer systems to maintain the pH of reaction mixtures in biochemical assays, electrophoresis, and enzyme catalysis studies. The high-purity formulation ensures minimal impurities and excellent performance in sensitive experiments.