Learn More
Alkali Scientific TRIS Hydrochloride [Tris(hydroxymethyl) aminomethane HCl], 500 G
SDP
Catalog No. 5000659762
Encompass_Preferred
TRIS Hydrochloride (Tris(hydroxymethyl) aminomethane HCl) is an essential biochemical reagent used for a variety of laboratory applications. The 500g package offers a moderate supply for most research labs engaged in molecular biology, biochemistry, and enzyme assays. TRIS Hydrochloride plays a crucial role in buffer preparations for experiments such as PCR, protein electrophoresis, and DNA/RNA extraction. Its ability to stabilize the pH of solutions makes it indispensable for maintaining optimal conditions for enzymatic reactions. This reagent is also commonly used in the formulation of TAE and TBE buffers for agarose gel electrophoresis and in various other biochemistry experiments.
Catalog No. 50-006-59762 Supplier Alkali Scientific Supplier No. ISH006CP
TRIS Hydrochloride (Tris(hydroxymethyl) aminomethane HCl) is an essential biochemical reagent used for a variety of laboratory applications. The 500g package offers a moderate supply for most research labs engaged in molecular biology, biochemistry, and enzyme assays. TRIS Hydrochloride plays a crucial role in buffer preparations for experiments such as PCR, protein electrophoresis, and DNA/RNA extraction. Its ability to stabilize the pH of solutions makes it indispensable for maintaining optimal conditions for enzymatic reactions. This reagent is also commonly used in the formulation of TAE and TBE buffers for agarose gel electrophoresis and in various other biochemistry experiments.