Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Hydrochloride, Ultra Pure, MP Biomedicals™

Catalog No. MP04816116 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04816116 100 g
MP04816124 500 g
MP04816100 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP04816116 Supplier MP Biomedicals, Inc Supplier No. 04816116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris-HCl is excellent biochemical and biological buffer for all applications where ultra high purity is required.

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per°C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

For Use With (Application) Molecular Biology
Physical Form Crystalline Powder
Quantity 100 g
Solution Type Buffer
Solubility Information Solubility (5% aq soln) Clear, Colorless Solution.
Formula Weight 157.6g/mol
Percent Purity ≥99%
Grade Ultra Pure
Recommended Storage Store at Room Temperature (15–30°C). Desiccate.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.