Learn More
Crescent Chemical Co Inc TRIS(HYDROXYMETHYL)AMINOMETHAN
SDP
Catalog No. 50248477
Encompass_Preferred
Tris(hydroxymethyl)aminomethane.hydrochloride molecular biology grade DNase/RNase Protease not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39787.01 CAS 1185-53-1
Catalog No. 50-248-477 Supplier Crescent Chemical Co Inc Supplier No. SE39787.01
Add to Cart
Add to Cart
Tris(hydroxymethyl)aminomethane.hydrochloride molecular biology grade DNase/RNase Protease not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39787.01 CAS 1185-53-1