Learn More
Crescent Chemical Co Inc TRIS(HYDROXYMETHYL)AMINOMETHAN
Supplier: Crescent Chemical Co Inc SE37180.02
Tris(hydroxymethyl)aminomethane analytical grade, USP Buffer quality for biological and enzymatic research. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 37180.02 CAS 77-86-1