Learn More
Crescent Chemical Co Inc TRIS(HYDROXYMETHYL)AMINOMETHAN
SDP
Supplier:  Crescent Chemical Co Inc SE37186.03
Encompass_Preferred
Tris(hydroxymethyl)aminomethane molecular biology grade DNase, RNase, Protease not detected. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 37186.03 CAS 77-86-1
Catalog No. 50-248-231