Learn More
Medchemexpress LLC Tris(hydroxymethyl)aminomethane | 77-86-1 | 99.9% | 500 G
SDP
Catalog No. 5000410711
Encompass_Preferred
THAM (Tris) is a biologically inert amino alcohol with low toxicity that buffers carbon dioxide and acids both in vitro and in vivo. It is an effective amine compound for pH control in the physiological range. It has been shown to reduce arterial carbon dioxide tension (PaCO2) levels and increase pH above 7.4 during infusion in pigs.
  • Biologically inert amino alcohol with low toxicity
  • Buffers carbon dioxide and acids
  • Effective for pH control in the physiological range
  • Reduces arterial carbon dioxide tension
  • Increases pH during infusion
Catalog No. 50-004-10711 Supplier Medchemexpress LLC Supplier No. HYD0227500G
THAM (Tris) is a biologically inert amino alcohol with low toxicity that buffers carbon dioxide and acids both in vitro and in vivo. It is an effective amine compound for pH control in the physiological range. It has been shown to reduce arterial carbon dioxide tension (PaCO2) levels and increase pH above 7.4 during infusion in pigs.
  • Biologically inert amino alcohol with low toxicity
  • Buffers carbon dioxide and acids
  • Effective for pH control in the physiological range
  • Reduces arterial carbon dioxide tension
  • Increases pH during infusion