Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry

Catalog No. AC140500025
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140500025 2.5 kg Plastic bottle
AC140505000 500 g Plastic bottle
AC140500010 1 kg Plastic bottle
AC140500100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC140500025 Supplier Thermo Scientific Chemicals Supplier No. 140500025
  • $334.00
    $279.65 / Each of 1

    Save $54.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg | CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance (40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
Melting Point >168.5°C
Color White
pH 10.0 to 11.5 (5% aq. soln.)
Boiling Point 219.0°C to 220.0°C (10.0 mmHg)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C4H11NO3
Linear Formula (HOCH2)3CNH2
Quantity 2.5 kg
Beilstein 04, 303
Merck Index 15, 9951
Solubility Information Solubility in water: 550g/L (25°C). Other solubilities: 79.1g/L ethylene glycol- 26g/L methanol (25°C), 22g/L 95% ethanol- 14g/L dmf (25°C), 2g/L acetone- 0.5g/L ethyl acetate (25°C), 0.05g/L chloroform- <0.05g/L ccl4 (25°C)
Molecular Weight (g/mol) 121.14
Formula Weight 121.14
Percent Purity 99+%
Grade Biochemistry
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
EINECSNumber 201-064-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.