Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)aminomethane, ACS Reagent (Tris; THAM; 2-Amino-2-(hydroxymethyl)-1,3-propanediol), Reagents

Catalog No. 01337637
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-637 100 g
01-337-636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-637 Supplier Reagents Holdings Llc Supplier No. C1342800100C2
 Restricted Item
Add to Cart
Add to Cart

Tris(hydroxymethyl)aminomethane, ACS Reagent (Tris; THAM; 2-Amino-2-(hydroxymethyl)-1,3-propanediol), ACS, use for Buffers, Acid Titrations. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tromethamine
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO

Specifications

Melting Point 171°C to 172°C
Boiling Point 219°C
Color White
For Use With (Equipment) pH Meter
Physical Form Solid
Quantity 100 g
Packaging Glass Bottle
Specific Gravity 1.35
Sterility Non-sterile
Grade ACS
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.