Learn More
Medchemexpress LLC Tris(hydroxymethyl)aminomethane-d11 (for molecular biology) | 202656-13-1 | 99.9% | 50 MG
SDP
Catalog No. 5000410283
Encompass_Preferred
THAM-d11 (for molecular biology) is the deuterium labeled Trometamol(Tris), for molecular biology. This compound is suitable for use as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-10283 Supplier Medchemexpress LLC Supplier No. HYD0227ES50MG
Add to Cart
Add to Cart
THAM-d11 (for molecular biology) is the deuterium labeled Trometamol(Tris), for molecular biology. This compound is suitable for use as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS