Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%

Catalog No. AAJ6256922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6256922 100 g
AAJ6256936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6256922 Supplier Thermo Scientific™ Supplier No. J6256922
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Used as a biological buffer substance in the pH range 7—9; pKa = 8.3 at 20°C; pKa = 7.82 at 37°C. Used as a biological buffer or as a component of buffer formulations such as TAE and TBE buffers. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms (pH 7.0-9.0). Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins. Used in chemical manufacturing (surface-active agents, vulcanization accelerators, and pharmaceuticals), as an emulsifying agent (cosmetics, mineral oil and paraffin wax emulsions, leather dressings, textile specialties, polishes, cleaning compounds, and soluble oils), and as an absorbent for acidic gases, biological buffer, acidimetric standard, and therapeutic alkalinizing agent. Used in buffer systems for the freezer storage of sperm, to regulate pH in the transport of live aquatic species, cell culture media, and anti-inflammatory drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound widely used as a biological buffer substance in the pH range 7—9; pKa = 8.3 at 20°C; pKa = 7.82 at 37°C. It is widely used as a biological buffer or as a component of buffer formulations such as TAE and TBE buffers. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms (pH 7.0-9.0). Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins. In medicine, Tris is used as alternative to sodium bicarbonate in the treatment of metabolic acidosis. Used in chemical manufacturing (surface-active agents, vulcanization accelerators, and pharmaceuticals), as an emulsifying agent (cosmetics, mineral oil and paraffin wax emulsions, leather dressings, textile specialties, polishes, cleaning compounds, and soluble oils), and as an absorbent for acidic gases, biological buffer, acidimetric standard, and therapeutic alkalinizing agent. Also used in buffer systems for the freezer storage of sperm, to regulate pH in the transport of live aquatic species, cell culture media, and anti-inflammatory drugs.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-hydroxymethyl-1,3-propanediol; TRIS
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168°C to 172°C
Boiling Point 219°C to 220°C (10mmHg)
Color White
Density 1.353
Physical Form Crystalline Powder
Quantity 100 g
Beilstein 741883
Merck Index 14,9772
Solubility Information Not miscible or difficult to mix in water. Soluble in ethylene glycol (79.1mg/ml),methanol (26mg/ml),anhydrous ethanol (14.6mg/ml),95% ethanol (22mg/ml),DMF (14mg/ml),acetone (2mg/ml),ethyl acetate (0.5mg/ml),olive oil (0.4mg/ml),chloroform (0.05mg/ml),and alkaline solution.
Formula Weight 121.14
Percent Purity 99.9%
Grade Electrophoresis
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-064-4
RTECSNumber TY2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.