Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 8.0, RNase free

Catalog No. AAJ60080AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60080AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60080AK Supplier Thermo Scientific Chemicals Supplier No. J60080AK
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Tris is used in pH control in buffering systems for electrophoresis applications.Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions. Also used as buffers in a wide variety of biological systems. It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color Colorless
Concentration 1M soln.
Solubility Information Fully miscible with water. Soluble in dimethyl sulfoxide and methanol.
pH 8.0
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride, RNAse free
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.