Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)aminomethane 99.0+%, TCI America™
SDP

Catalog No. A0321500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0321500G 500 g
A032125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0321500G Supplier TCI America Supplier No. A0321500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trimethylolaminomethane, Tris-Base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 170°C
Boiling Point 220°C
Color White-Yellow
For Use With (Application) Molecular Biology
pH 10.0 to 10.8 (0.1 mol/L aq 25°C)
Physical Form Crystals or Powder
Quantity 500 g
Formula Weight 121.14
Percent Purity ≥99.0% (T)
Grade Laboratory
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
EINECSNumber (2)-0318
RTECSNumber TY2900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.