Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)aminomethane, USP, EP, bioCERTIFIED, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18614851 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
12 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-851 1 kg Plastic Bottle
18-614-852 2.5 kg Plastic Bottle
18-614-853 12 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-614-851 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T97001KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO

Specifications

Color Clear/Colorless
pH 10 to 11.5
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range 168-172°C
Packaging Plastic Bottle
Quantity 1 kg
Residual Solvents To pass test
Percent Purity 99 to 100.5%
Grade USP, EP, bioCERTIFIED
Related Substances To pass test
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.