Learn More
Crescent Chemical Co Inc TRIS(HYDROXYMETHYL)METHYLGLYCI
SDP
Catalog No. 50248242
Encompass_Preferred
Tris(hydroxymethyl)methylglycine analytical grade MP 179 - 187 °C; pKa 20 = 8.15. Buffering substance (1). Non-toxic substitute for barbital (2). Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 37195.03 CAS 1389-47-5
Catalog No. 50-248-242 Supplier Crescent Chemical Co Inc Supplier No. SE37195.03
Tris(hydroxymethyl)methylglycine analytical grade MP 179 - 187 °C; pKa 20 = 8.15. Buffering substance (1). Non-toxic substitute for barbital (2). Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 37195.03 CAS 1389-47-5