Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)phosphine, 95%

Catalog No. p-3737703
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363840010 1 g Glass bottle
AC363840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC363840010 Supplier Thermo Scientific Chemicals Supplier No. 363840010
Only null left
Add to Cart

CAS: 2767-80-8 | C3H9O3P | 124.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2767-80-8
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.08
MDL Number MFCD00055382
InChI Key JMXMXKRNIYCNRV-UHFFFAOYSA-N
Synonym tris hydroxymethyl phosphine, methanol, phosphinidynetris, trimethylolphosphine, phosphinidynetrimethanol, tris methanol phosphine, phosphinidynetrismethanol, unii-y6tg7wf7oq, y6tg7wf7oq, methanol, 1,1',1-phosphinidynetris, methanol, phosphinidynetri-7ci,8ci
PubChem CID 76001
IUPAC Name bis(hydroxymethyl)phosphanylmethanol
SMILES C(O)P(CO)CO

Specifications

Melting Point 48°C to 56°C
Color White to Yellow
Boiling Point 111.0°C to 113.0°C (2.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 124.08
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Tris(hydroxymethyl)phosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 220-445-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.