Learn More
Strem, An Ascensus Company CAS# 4706-17-6. 10g. Tris(3-hydroxypropyl)phosphine, min. 84%. MFCD00269835
SDP
Supplier:  Strem, An Ascensus Company 15637510G
Encompass
CAS# 4706-17-6. 10g. Tris(3-hydroxypropyl)phosphine, min. 84%. MFCD00269835. Molecular Weight: 208.24. Molecular Formula: (HOC3H6)3P. Color/form: viscous liq. Strem# 15-6375. http://www.strem.com/catalog/v/15-6375/
Catalog No. 50-901-10424