Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Hydroxypropyl)Phosphine

Catalog No. 7778541G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-785-41G 5 g
77-785-45G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 77-785-41G Supplier Sigma Aldrich Supplier No. 7778541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(hydroxypropyl)phosphine can be used as a:Reducing agent for the cleavage of disulfide bonds in both aqueous and buffered aqueous-organic medium.Ligand in the synthesis of dendritic macromolecules which are used as catalysts in heterogenizing homogeneous reactions.Reducing agent for the synthesis of disulfide cyclopeptides via one-pot peptide ligation-oxidative cyclization.

Specifications

CAS 4706-17-6
Density 1.136 g/mL at 25°C
Molecular Formula C9H21O3P
Linear Formula C9H21O3P
MDL Number MFCD00269835
Quantity 5 g
Synonym THPP
Molecular Weight (g/mol) 208.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.