Learn More
Medchemexpress LLC Tris (maleate) | 72200-76-1 | MFCD00082442 | 98.0% | 237.21 g·mol^-1 | C8H15NO7 | 50 G
SDP
Catalog No. 5000285219
Encompass_Preferred
TRIS (maleate) is the maleate salt of tris(hydroxymethyl)aminomethane, supplied as a high-purity white crystalline powder for use as a buffering reagent in biological and biochemical research. It is commonly used to adjust pH and ionic strength in growth media, tissue preparations, and as a diluent or washing buffer; handle and store according to the product safety data sheet.
  • High purity (98.0%) suitable for research use.
  • Effective buffering capacity for biological media and assays.
  • Compatible with growth media, tissue preparations, and washing steps.
  • White crystalline powder form for accurate weighing and dissolution.
  • Store sealed at 4°C; follow frozen storage recommendations for solutions.
Catalog No. 50-002-85219 Supplier Medchemexpress LLC Supplier No. HYW39337250G
TRIS (maleate) is the maleate salt of tris(hydroxymethyl)aminomethane, supplied as a high-purity white crystalline powder for use as a buffering reagent in biological and biochemical research. It is commonly used to adjust pH and ionic strength in growth media, tissue preparations, and as a diluent or washing buffer; handle and store according to the product safety data sheet.
  • High purity (98.0%) suitable for research use.
  • Effective buffering capacity for biological media and assays.
  • Compatible with growth media, tissue preparations, and washing steps.
  • White crystalline powder form for accurate weighing and dissolution.
  • Store sealed at 4°C; follow frozen storage recommendations for solutions.