Learn More
Medchemexpress LLC Tris (maleate) | 72200-76-1 | MFCD00082442 | 98.0% | 237.21 g/mol | C8H15NO7 | 10 G
SDP
Catalog No. 5000285039
Encompass_Preferred
Tris (maleate) is the maleate salt of tris(hydroxymethyl)aminomethane used as a buffering agent in biochemical and cell biology applications. It stabilizes pH in growth media, tissue preparations, and washing or diluent solutions, and is supplied as a highly soluble reagent to support reproducible experimental workflows.
  • Buffered pH support for media and tissue preparations.
  • Water-soluble; also soluble in DMSO with sonication if needed.
  • High purity: 98.0%.
  • Suitable for use as a diluent or washing buffer.
  • Available in laboratory-scale package sizes such as 10 G.
Catalog No. 50-002-85039 Supplier Medchemexpress LLC Supplier No. HYW39337210G
Tris (maleate) is the maleate salt of tris(hydroxymethyl)aminomethane used as a buffering agent in biochemical and cell biology applications. It stabilizes pH in growth media, tissue preparations, and washing or diluent solutions, and is supplied as a highly soluble reagent to support reproducible experimental workflows.
  • Buffered pH support for media and tissue preparations.
  • Water-soluble; also soluble in DMSO with sonication if needed.
  • High purity: 98.0%.
  • Suitable for use as a diluent or washing buffer.
  • Available in laboratory-scale package sizes such as 10 G.