Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tris-maleate, 98%

Catalog No. AC264971000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264971000 100 g Glass bottle
AC264970250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC264971000 Supplier Thermo Scientific Chemicals Supplier No. 264971000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 72200-76-1 | C4H11NO3·C4H4O4 | 237.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72200-76-1
Molecular Formula C4H11NO3·C4H4O4
Molecular Weight (g/mol) 237.21
MDL Number MFCD00082442
InChI Key HTMWOUBCEZXSHN-BTJKTKAUSA-N
Synonym butenedioic acid; tris buffer, 2-amino-2-hydroxymethyl propane-1,3-diol; but-2-enedioic acid
PubChem CID 16218780
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;(Z)-but-2-enedioic acid
SMILES C(C(CO)(CO)N)O.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 104°C to 106°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: 1g/10 mL
Formula Weight 237.21
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tris-maleate, 98%
Show More Show Less
EINECSNumber 276-455-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.