Learn More
Electron Microscopy Sciences Tris Maleate Buffer 0.2M, pH range 6.4-8.0 250 ML
SDP
Catalog No. 50980237
Encompass_Preferred
Please specify pH when ordering.
pH: 6.4-8.0
Catalog No. 50-980-237 Supplier Electron Microscopy Sciences Supplier No. 11730
Please specify pH when ordering.
pH: 6.4-8.0