Learn More
Strem, An Ascensus Company CAS# 329735-72-0. 1g. Tris(methylcyclopentadienyl)yttrium (99.9%-Y) (REO). MFCD00145485
SDP
Supplier:  Strem, An Ascensus Company 3950501G
Encompass
CAS# 329735-72-0. 1g. Tris(methylcyclopentadienyl)yttrium (99.9%-Y) (REO). MFCD00145485. Molecular Weight: 326.27. Molecular Formula: (CH3C5H4)3Y. Color/form: yellow xtl. Strem# 39-5050. http://www.strem.com/catalog/v/39-5050/
Catalog No. 50-204-5299