Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS, MP Biomedicals™

Catalog No. MP04819623 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04819623 1 kg
MP04819620 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04819623 Supplier MP Biomedicals, Inc Supplier No. 04819623
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TRIS Ultra Pure, ≥99.9%, White crystalline powder

  • Tris is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
  • Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions.
  • Tris have been useful as buffers in a wide variety of biological systems.
  • It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
  • It does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
  • It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
  • It is RNAse and DNAse-free.Tris is relatively non-hygroscopic;but, if needed, it can be dried at 100°C for up to 4 hours to remove any water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris-[hydroxymethyl]aminomethane, THAM, 2-Amino-2-(hydroxymethyl)- 1,3-propandiol, Tromethamine, Trometamol

Specifications

Melting Point 165°C to 175°C
Boiling Point 219°C to 220°C (at 10 mmHg)
Color White
For Use With (Application) Electrophoresis
pH 10 to 11.5 (5% aq soln)
Physical Form Crystalline Powder
Quantity 1 kg
Solution Type Buffer
Loss on Drying ≤1%
Beilstein 741883
Solubility Information Soluble in water (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL)
Percent Purity ≥99%
Grade ≥99.9%
Show More Show Less
RTECSNumber TY2900000
storageNote1 Room Temperature, desiccate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.