Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS, MP Biomedicals™

Catalog No. MP1TRIS05KG Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1TRIS05KG 5 kg
MP1TRIS0500 500 g
MP1TRIS01KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP1TRIS05KG Supplier MP Biomedicals, Inc Supplier No. 11TRIS05KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TRIS, Molecular biology grade, >99.8%, White Crystalline Powder

  • Tris may be used by itself as a buffer or as a component of mixed buffer formulations e.g. reaction buffers for restriction enzymes or nucleic acid modifying enzymes.
  • Tris salts are used in protein crystallization at various pH values.
  • The use of low-ionic strength Tris buffers in the formation of intermediate filaments of lamin from Caenorhabditis elegans has been described.
  • Tris has been utilized in studies of double stranded complexes of peptide nucleic acids (PNA) and their complementary DNA sequences.
  • The use of Tris in capillary electrochromatography and UV analysis of tocopherols and tocotrienols has been reported.
  • Tris is an established basimetric standard and buffer used in biochemistry and molecular biology.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris-(hydroxymethyl) aminomethane, Trometamol

Specifications

Melting Point 167°C to 172°C (literature)
Boiling Point 219°C to 220°C/10 mmHg(literature)
Color White
pH 10.5 to 11.5 (0.5 M aq. soln., 20°C)
Physical Form Crystalline Powder
Quantity 5 kg
Solution Type Buffer
Beilstein 741883
Solubility Information Soluble in water (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), chloroform (0.05mg/mL), and alkaline solution.
Percent Purity >99.8%
Grade >99.8%
Show More Show Less
RTECSNumber TY2900000
storageNote1 Store at Room temperature(15-30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.