Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris, MP Biomedicals™

Catalog No. MP21031335 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21031335 500 g
MP21031331 100 g
MP21031332 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP21031335 Supplier MP Biomedicals, Inc Supplier No. 02103133.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Useful as a buffer in a variety of biological systems. Tris, MP Biomedicals is used for pH control of body fluids and in buffering systems for electrophoresis. It is ultra-pure, RNAse and DNAse-free, and relatively non-hygroscopic. It can be dried at 100°

Tris have been useful as buffers in a wide variety of biological systems. It is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.

  • Used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions
  • Used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes)
  • Does not precipitate calcium salts and is of value in maintaining solubility of manganese salts
  • Can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol, Tromethamine, Tris-[hydroxymethyl]amino-methane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

For Use With (Application) Electrophoresis
pH 10.4
Physical Form Crystalline Powder
Quantity 500 g
Solution Type Buffer
Solubility Information Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL).
Formula Weight 121.14
Percent Purity >99.9%
Grade Ultrapure
Recommended Storage Store at Room Temperature (15–30°C). Store Desiccated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.