Learn More
Strem, An Ascensus Company CAS# 4731-65-1. 1g. Tris(o-methoxyphenyl)phosphine, min. 98%. MFCD00014892
SDP
Catalog No. 5090110520
Encompass
CAS# 4731-65-1. 1g. Tris(o-methoxyphenyl)phosphine, min. 98%. MFCD00014892. Molecular Weight: 352.37. Molecular Formula: (o-CH3OC6H4)3P. Color/form: white xtl. Strem# 15-7950. http://www.strem.com/catalog/v/15-7950/
Catalog No. 50-901-10520 Supplier Strem, An Ascensus Company Supplier No. 1579501G
CAS# 4731-65-1. 1g. Tris(o-methoxyphenyl)phosphine, min. 98%. MFCD00014892. Molecular Weight: 352.37. Molecular Formula: (o-CH3OC6H4)3P. Color/form: white xtl. Strem# 15-7950. http://www.strem.com/catalog/v/15-7950/