Learn More
Strem, An Ascensus Company CAS# 18437-78-0. 5g. Tris(p-fluorophenyl)phosphine, 99%. MFCD00013553
SDP
Catalog No. 5090110512
Encompass
CAS# 18437-78-0. 5g. Tris(p-fluorophenyl)phosphine, 99%. MFCD00013553. Molecular Weight: 316.27. Molecular Formula: (p-FC6H4)3P. Color/form: off-white xtl. Strem# 15-7890. http://www.strem.com/catalog/v/15-7890/
Catalog No. 50-901-10512 Supplier Strem, An Ascensus Company Supplier No. 1578905G
CAS# 18437-78-0. 5g. Tris(p-fluorophenyl)phosphine, 99%. MFCD00013553. Molecular Weight: 316.27. Molecular Formula: (p-FC6H4)3P. Color/form: off-white xtl. Strem# 15-7890. http://www.strem.com/catalog/v/15-7890/