Learn More
Strem, An Ascensus Company CAS# 1109-15-5. 5g. Tris(pentafluorophenyl)borane, min. 97%. MFCD00269813
SDP
Catalog No. 5090108766
Encompass
CAS# 1109-15-5. 5g. Tris(pentafluorophenyl)borane, min. 97%. MFCD00269813. Molecular Weight: 511.99. Molecular Formula: (C6F5)3B. Color/form: off-white pwdr. Strem# 05-1500. http://www.strem.com/catalog/v/05-1500/
Catalog No. 50-901-08766 Supplier Strem, An Ascensus Company Supplier No. 0515005G
CAS# 1109-15-5. 5g. Tris(pentafluorophenyl)borane, min. 97%. MFCD00269813. Molecular Weight: 511.99. Molecular Formula: (C6F5)3B. Color/form: off-white pwdr. Strem# 05-1500. http://www.strem.com/catalog/v/05-1500/