Learn More
Bioworld PEB- Protein Extraction Buffer (5x), 250 mL
Catalog No. 50253061
Encompass_Preferred
Tris Protein Extraction Buffer (PEB), 5X Lysis buffer for extracting proteins from cells. Technical Information:5X concentrationTris-HCL, pH 8.0 (0.5M)B-mercaptoethanol (100mM)EDTA (20mM)Add protease inhibitor according to your protocol
Catalog No. 50-253-061 Supplier Bioworld Supplier No. 220400561
Tris Protein Extraction Buffer (PEB), 5X Lysis buffer for extracting proteins from cells. Technical Information:5X concentrationTris-HCL, pH 8.0 (0.5M)B-mercaptoethanol (100mM)EDTA (20mM)Add protease inhibitor according to your protocol