Learn More
PROVISION KINETICS INC TRIS- SALINE BUFFER
Catalog No. NC0493545
Encompass
500 ml bottle of Tris-saline buffer, pH 8.4 designed for use with Anti-IIa and Anti-Xa heparin test kits (sold separately)
Catalog No. NC0493545 Supplier PROVISION KINETICS INC Supplier No. BUF500
500 ml bottle of Tris-saline buffer, pH 8.4 designed for use with Anti-IIa and Anti-Xa heparin test kits (sold separately)