Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(tetrathiafulvalene) Bis(tetrafluoroborate) Complex, TCI America™
SDP

Catalog No. T12821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12821G Supplier TCI America Supplier No. T12821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55492-86-9
Molecular Formula C18H12B2F8S12
Molecular Weight (g/mol) 786.62
MDL Number MFCD00145489
InChI Key IJZRAQUJMWAFMU-UHFFFAOYSA-N
Synonym Tetrathiafulvalene Fluoroborate
PubChem CID 102601231
IUPAC Name 2-(1,3-dithiol-1-ium-2-yl)-1,3-dithiol-1-ium;2-(1,3-dithiol-2-ylidene)-1,3-dithiole;ditetrafluoroborate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CSC(=C2SC=CS2)S1.C1=CSC(=C2SC=CS2)S1.C1=C[S+]=C(S1)C2=[S+]C=CS2

Specifications

Color Black
Quantity 1 g
UN Number 1759
Formula Weight 786.62
Physical Form Crystalline Powder
Chemical Name or Material Tris(tetrathiafulvalene) Bis(tetrafluoroborate) Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.