Learn More
Research Products International Corp TRIS-Tricine-SDS Running Buffer 10X Solution, 1 Liter
SDP
Catalog No. 50997991
Encompass_Preferred
10X Formulation:
TRIS: 1 M
Tricine: 1 M
SDS (pH 8.3): 1%
Catalog No. 50-997-991 Supplier Research Products International Corp Supplier No. T273001000.0
10X Formulation:
TRIS: 1 M
Tricine: 1 M
SDS (pH 8.3): 1%