Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8.3

Catalog No. p-7076556
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60992AP 500 mL
AAJ60992K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60992AP Supplier Thermo Scientific Chemicals Supplier No. J60992AP
Only null left
Add to Cart

Tris Tricine Gel Electrophoresis is routinely used for the separation of small proteins and peptides with a molecular weight of I to 100 kDa. It is used for protein analysis. This type of gel is perfectly suited for the separation and identification of proteins in samples containing only a few different proteins to samples containing complex mixtures (e.g. from cell extracts).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TRIS-Tricine-SDS running buffer (10X)
Physical Form Liquid
Quantity 500 mL
Packaging Plastic bottle
Solubility Information It is soluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.