Learn More
Alkali Scientific TRIS Tricine SDS Running Buffer 10X Solution, 500 Milliliters
SDP
Catalog No. 5000660184
Encompass_Preferred
TRIS Tricine SDS Running Buffer 10X Solution is a highly concentrated electrophoresis buffer designed to improve protein separation during SDS-PAGE. This 500mL bottle of 10X solution can be diluted to make multiple liters of running buffer for electrophoresis, reducing the need for frequent reorders. Tricine helps achieve better resolution for small molecular weight proteins, making this buffer ideal for detailed protein profiling. This solution stabilizes proteins during electrophoresis and ensures efficient migration through gels, delivering high-quality results in Western blotting and other protein analysis techniques commonly used in molecular biology and biochemistry laboratories.
Catalog No. 50-006-60184 Supplier Alkali Scientific Supplier No. ESD002CP
TRIS Tricine SDS Running Buffer 10X Solution is a highly concentrated electrophoresis buffer designed to improve protein separation during SDS-PAGE. This 500mL bottle of 10X solution can be diluted to make multiple liters of running buffer for electrophoresis, reducing the need for frequent reorders. Tricine helps achieve better resolution for small molecular weight proteins, making this buffer ideal for detailed protein profiling. This solution stabilizes proteins during electrophoresis and ensures efficient migration through gels, delivering high-quality results in Western blotting and other protein analysis techniques commonly used in molecular biology and biochemistry laboratories.