Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trifluoro-2,4-pentanedionato)chromium(III) 98.0+%, TCI America™
SDP

Catalog No. T09321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T09321G Supplier TCI America Supplier No. T09321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14592-89-3
Molecular Formula C15H15CrF9O6
Molecular Weight (g/mol) 514.26
MDL Number MFCD00059456
InChI Key YPJYEKCGHXZCQF-RQQUGGEBSA-N
Synonym Chromium(III) Trifluoroacetylacetonate, Trifluoroacetylacetono Chromium(III) Salt

Specifications

Color Black-Gray
Quantity 1 g
Formula Weight 511.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(trifluoro-2,4-pentanedionato)chromium(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.