Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trifluoro-2,4-pentanedionato)iron(III) 98.0+%, TCI America™
SDP

Catalog No. T07505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07505G Supplier TCI America Supplier No. T07505G

Chemical Identifiers

CAS 28736-69-8
Molecular Formula C15H12F9FeO6
Molecular Weight (g/mol) 515.086
MDL Number MFCD00017760
InChI Key DYSQBHKDTAZKQW-VFEMKWSISA-K
Synonym Ferric(III) Trifluoroacetylacetonate, Iron(III) Trifluoroacetylacetonate, Trifluoroacetylacetono Iron(III) Salt
PubChem CID 53384603
IUPAC Name iron(3+);(E)-1,1,1-trifluoro-4-oxopent-2-en-2-olate
SMILES CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].[Fe+3]

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 515.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(trifluoro-2,4-pentanedionato)iron(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.