Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(trimethylsilyl) phosphite

Catalog No. 934125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-412-5ML 5 mL
93-412-100ML 100 mL
93-412-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 93-412-5ML Supplier Sigma Aldrich Supplier No. 934125ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction); Review

Specifications

CAS 1795-31-9
Density 0.893 g/mL (at 25°C (literature))
Boiling Point 78°C to 81°C (8 mmHg)
Molecular Formula C9H27O3PSi3
Refractive Index n20/D 1.409 (literature)
Linear Formula [(CH3)3 SiO]3 P
MDL Number MFCD00015588
Quantity 5 mL
Molecular Weight (g/mol) 298.54
Percent Purity ≥95.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.