Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Trimethylsilyl)Phosphine

Catalog No. 3336701G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-367-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 33-367-01G Supplier Sigma Aldrich Supplier No. 3336701G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(trimethylsilyl)phosphine (P(TMS)3) can be used as a precursor for the syntheses of:Indium phosphide (InP) and indiu mgallium phosphide (InGaP) nanocrystals, type III-V semiconductors.Chiral TMS-phospholane which can be further used to synthesize various bisphospholane ligands for asymmetric catalysis.

Specifications

CAS 31087-44-2
Density 0.809 g/mL at 20°C
Boiling Point 119°C to 120°C
Molecular Formula C5H12O
Linear Formula CH3CH2CH2CH(OH)CH3
Refractive Index n20/D 1.502
Quantity 1 g
MDL Number MFCD00065953
Synonym (P(TMS)3); Tris(trimethylsilyl)phosphane
Molecular Weight (g/mol) 88.15 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.