Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyl)phosphine (ca. 10% in Hexane), TCI America™
SDP

Catalog No. T345310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T345310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T345310G Supplier TCI America Supplier No. T345310G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15573-38-3
Molecular Formula C9H27PSi3
Molecular Weight (g/mol) 250.544
MDL Number MFCD00015487
InChI Key OUMZKMRZMVDEOF-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphine, tris trimethylsilyl phosphane, phosphine, tris trimethylsilyl, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane, tris trimethylsilyl phosphine in hexane, pubchem6456, acmc-1btki, tris-trimethylsilyl phosphine, phosphine,tris trimethylsilyl
PubChem CID 272683
IUPAC Name tris(trimethylsilyl)phosphane
SMILES C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C

Specifications

Color Yellow
Quantity 10 g
UN Number 1208
Formula Weight 250.54
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl)phosphine (ca. 10% in Hexane)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.