Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Trimethylsilyl)Silane

Catalog No. 36071625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-071-625G 25 g
36-071-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 36-071-625G Supplier Sigma Aldrich Supplier No. 36071625G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(trimethylsilyl)silane is an organosilicon generally used as a radical reducing agent for xanthates, organic halides, isocyanides, selenides, and acid chlorides. It is also used for hydrosilylation of alkenes, alkynes, and dialkyl ketones.Super Silyl Protecting GroupsUsed in:HydrosilylationsRadical reactionsReductions of acid chloridesReductions of carbon-halogen bondsHydrosilations of carbonylsAnother common application involves the use of the tris(trimethylsilyl)silyl (TTMSS, or super silyl) group when complexed with transition metals and main group elementsMore recently, the super silyl group is being utilized in carbon-carbon bond forming reactions

Specifications

CAS 1873-77-4
Density 0.806 g/mL at 25°C
Boiling Point 73°C (5 mmHg, lit.)
Molecular Formula C9H28Si4
Refractive Index n20/D 1.489
Linear Formula [(CH3)3Si]3SiH
MDL Number MFCD00077893
Quantity 25 g
Synonym 1,1,1,3,3,3-Hexamethyl-2-trimethylsilyl-trisilane; TTMSS
Molecular Weight (g/mol) 248.66 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.