Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Trimethylsilyl)Silanol

Catalog No. 9024891G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-248-91G 5 g
90-248-95G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-248-91G Supplier Sigma Aldrich Supplier No. 9024891G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Under a dual catalytic copper/photoredox manifold, this supersilanol has been demonstrated by the MacMillan lab to be an excellent reagent for the trifluoromethylation of alkyl halides and aryl halides to yield alkyl-CF3 and aryl-CF3 in high yields. In both cases, these reactions exhibit wide substrate scope with good functional group tolerance. More specifically, a variety of 5-membered and 6-membered heteroaryl halides can be readily converted to the corresponding trifluoromethylheteroarenes under mild conditions. To be use in conjunction with dMesSCF3 (901466) and Ir photocatalyst (902217 or 902225).A radical approach to the copper oxidative addition problem: Trifluoromethylation of bromoarenes

Specifications

CAS 7428-60-6
Density 0.859 g/mL
Molecular Formula C9H28OSi4
Refractive Index n/D 1.496
Linear Formula C9H28OSi4
MDL Number MFCD00093727
Quantity 5 g
Synonym (Hydroxy-bis(trimethylsilyl)silyl)-trimethylsilane; (TMS)3SiOH; 1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)trisilan-2-ol; Supersilanol
Molecular Weight (g/mol) 264.66 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.