Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(triphenylphosphine)ruthenium(II) Dichloride 90.0+%, TCI America™
SDP

Catalog No. D19975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19975G 5 g
D19971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19975G Supplier TCI America Supplier No. D19975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15529-49-4
Molecular Formula C54H45Cl2P3Ru
Molecular Weight (g/mol) 958.85
MDL Number MFCD00013077
InChI Key KSHWMFNJCNGTJO-UHFFFAOYSA-L
Synonym dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
PubChem CID 11007548
IUPAC Name λ¹-ruthenium(1+) tris(triphenylphosphane) dichloride
SMILES [Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Black-Green
Quantity 5 g
Formula Weight 958.85
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(triphenylphosphine)ruthenium(II) Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.