Learn More
Bioworld Tris-Triton Buffer, 500 mL
Catalog No. NC0847011
Encompass_Preferred
Tris-Triton bufferis the idealbuffer with detergent required to release membrane- or cytoskeleton- bound proteins.Contains: 10mM Tris (pH 7.4), 100mM NaCl, 1mM EDTA, 1mM EGTA, 1% Triton X-100, 10% Glycerol and 0.1% SDSAppearance: Liquid
Catalog No. NC0847011 Supplier Bioworld Supplier No. 420203101
Tris-Triton bufferis the idealbuffer with detergent required to release membrane- or cytoskeleton- bound proteins.Contains: 10mM Tris (pH 7.4), 100mM NaCl, 1mM EDTA, 1mM EGTA, 1% Triton X-100, 10% Glycerol and 0.1% SDSAppearance: Liquid