Learn More
National Diagnostics TRIS - ULTA PURE 1 KG
SDP
Catalog No. 5089990033
Encompass
Tris - ULTA PURE, Purified to remove ammonia and amine contaminants, 1 KG
Catalog No. 50-899-90033 Supplier National Diagnostics Supplier No. EC4061KG
Tris - ULTA PURE, Purified to remove ammonia and amine contaminants, 1 KG