Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris, Ultra Pure, 5kg, MP Biomedicals

Catalog No. ICN819638 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN819638 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN819638 Supplier MP Biomedicals, Inc Supplier No. 04819638
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

77-86-1 Purity: 99.9% Heavy metals (as Pb): <<5 ppm O.D. (260mm) of a 5% saturated solution: <<0.1 pKa at 20°C = 8.1 Biological buffer reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

For Use With (Application) Electrophoresis
Physical Form Powder
Quantity 5 kg
Solution Type Buffer
Percent Purity ≥99%
Grade Ultrapure
Chemical Name or Material Tris
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.