Learn More
Strem, An Ascensus Company CAS# 370878-69-6. 250mg. Tris[5-fluoro-2-(2-pyridinyl-kN)phenyl-kC]iridium(III), 95%.
SDP
Supplier:  Strem, An Ascensus Company 776100250MG
Encompass
CAS# 370878-69-6. 250mg. Tris[5-fluoro-2-(2-pyridinyl-kN)phenyl-kC]iridium(III), 95%. . Molecular Weight: 708.75. Molecular Formula: C33H21F3IrN3. Color/form: yellow pwdr. Strem# 77-6100. http://www.strem.com/catalog/v/77-6100/
Catalog No. NC1591210