Learn More
Medchemexpress LLC Trisbenzyltriazolylm 250mg | 510758-28-8 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY116677250MG
Encompass_Preferred
Tris(benzyltriazolylmethyl)amine (TBTA) is a tridentate ligand used to stabilize copper(I) catalysts in copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions. It is employed to improve efficiency and selectivity in bioconjugation workflows for tagging proteins, peptides, and small molecules.
  • Stabilizes Cu(I) to improve click reaction efficiency.
  • Enhances yield and selectivity in CuAAC conjugations.
  • Suitable for protein, peptide, and small-molecule labeling.
  • Available as solid and DMSO solution formats for flexible use.
  • High purity supports reproducible experimental results.
Catalog No. 50-002-27143