Learn More
Medchemexpress LLC HY-116677 50mg Medchemexpress, Tris(benzyltriazolylmethyl)amine CAS:510758-28-8 Purity:>98%
SDP
Catalog No. 502080894
Encompass_Preferred
Medchemexpress, HY-116677 50mg Tris(benzyltriazolylmethyl)amine CAS:510758-28-8 Tris(benzyltriazolylmethyl)amine (TBTA) is a ligand that acts as a biochemical tool for the tagging of proteins and enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0894 Supplier Medchemexpress LLC Supplier No. HY11667750MG
Medchemexpress, HY-116677 50mg Tris(benzyltriazolylmethyl)amine CAS:510758-28-8 Tris(benzyltriazolylmethyl)amine (TBTA) is a ligand that acts as a biochemical tool for the tagging of proteins and enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.